Anti-COLQ Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP34362_P050 ARP34362_T100
AVIVARP34362_P050EA 401 EUR
AVIVARP34362_P050 AVIVARP34362_T100
Anti-COLQ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: COLQ (collagen-like tail subunit (single strand of homotrimer) of asymmetric acetylcholinesterase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR