Anti-ZNF575 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP34490_P050EA 385 EUR
AVIVARP34490_P050
Anti-ZNF575 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF575 (zinc finger protein 575)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR