Anti-ZNF568 Rabbit Polyclonal Antibody
Supplier: Aviva Systems Biology
AVIVARP34876_T100EA 335 EUR
AVIVARP34876_T100
Anti-ZNF568 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF568 (zinc finger protein 568)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now