Anti-CLIC1 Rabbit Polyclonal Antibody
Supplier: Aviva Systems Biology
AVIVARP35044_P050EA 385 EUR
AVIVARP35044_P050 AVIVARP35044_T100
Anti-CLIC1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLIC1 (chloride intracellular channel 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now