Anti-KCNJ3 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP35081_P050EA 385 EUR
AVIVARP35081_P050
Anti-KCNJ3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KCNJ3 (potassium inwardly-rectifying channel, subfamily J, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Guinea pig, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR