Anti-GABRB2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP35338_T100EA 335 EUR
AVIVARP35338_T100
Anti-GABRB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GABRB2 (gamma-aminobutyric acid (GABA) A receptor, beta 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR