Anti-THRB Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP35635_P050EA 385 EUR
AVIVARP35635_P050
Anti-THRB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: THRB (thyroid hormone receptor, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR