Anti-SLC18A1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP35649_P050EA 385 EUR
AVIVARP35649_P050
Anti-SLC18A1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC18A1 (solute carrier family 18 (vesicular monoamine transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR