Anti-ZNF317 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP39460_P050EA 385 EUR
AVIVARP39460_P050
Anti-ZNF317 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF317 (zinc finger protein 317)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR