Anti-HOXB4 Rabbit Polyclonal Antibody
Supplier: Aviva Systems Biology
ARP39581_P050 ARP39581_T100
AVIVARP39581_P050EA 401 EUR
AVIVARP39581_P050 AVIVARP39581_T100
Anti-HOXB4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HOXB4 (homeobox B4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now