Anti-SLC16A8 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP43935_T100EA 335 EUR
AVIVARP43935_T100
Anti-SLC16A8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC16A8 (solute carrier family 16 (monocarboxylate transporter), member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR