Anti-LRRC15 Rabbit Polyclonal Antibody
Supplier: Aviva Systems Biology
AVIVARP50292_P050EA 385 EUR
AVIVARP50292_P050
Anti-LRRC15 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LRRC15 (leucine rich repeat containing 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now