Anti-TREM1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP63526_P050EA 385 EUR
AVIVARP63526_P050
Anti-TREM1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TREM1 (triggering receptor expressed on myeloid cells 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR