Anti-MGME1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP69139_P050EA 385 EUR
AVIVARP69139_P050
Anti-MGME1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MGME1 (mitochondrial genome maintenance exonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR