Anti-TNK1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAM1861BEA 348 EUR
ABGEAM1861B
Anti-TNK1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TNK1 (tyrosine kinase, non-receptor, 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR