Anti-CCNJL Rabbit Polyclonal Antibody [clone: 170CT7.5.4]
Supplier: Abgent
ABGEAP11307CEA 348 EUR
ABGEAP11307C
Anti-CCNJL Rabbit Polyclonal Antibody [clone: 170CT7.5.4]
Antibodies
Type: Primary
Antigen: CCNJL (cyclin J-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human