Anti-COQ6 Rabbit Polyclonal Antibody [clone: 275CT1]

Supplier: Abgent
ABGEAP11456AEA 348 EUR
ABGEAP11456A
Anti-COQ6 Rabbit Polyclonal Antibody [clone: 275CT1]
Antibodies
Type: Primary
Antigen: COQ6 (coenzyme Q6 monooxygenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR