Anti-SMC1A Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP13544AEA 348 EUR
ABGEAP13544A
Anti-SMC1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMC1A (structural maintenance of chromosomes 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR