Anti-SLC11A2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP13801CEA 348 EUR
ABGEAP13801C
Anti-SLC11A2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC11A2 (solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR