Anti-GAPDH Goat polyclonal antibody

Supplier: OriGene
ORIGTA302944EA 547 EUR
ORIGTA302944
Anti-GAPDH Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 0.01-0.03?g/ml; Immunochemistry: 0.3?g/ml;

Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR