Anti-FGF21 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA303289EA 550 EUR
ORIGTA303289
Anti-FGF21 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: FGF21 (fibroblast growth factor 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR