Anti-PIK3R3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA308969EA 681 EUR
ORIGTA308969
Anti-PIK3R3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: PIK3R3 (phosphoinositide-3-kinase, regulatory subunit 3 (gamma))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR