Anti-NFKBIZ Mouse Monoclonal Antibody [clone: OTI1E8]
Supplier: OriGene
ORIGTA502768EA 639 EUR
ORIGTA502768
Anti-NFKBIZ Mouse Monoclonal Antibody [clone: OTI1E8]
Antibodies
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100
Type: Primary
Antigen: NFKBIZ (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human