Anti-MGLL Mouse Monoclonal Antibody [clone: OTI1B1]

Supplier: OriGene
ORIGTA503068EA 616 EUR
ORIGTA503068
Anti-MGLL Mouse Monoclonal Antibody [clone: OTI1B1]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MGLL (monoglyceride lipase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR