Anti-KCNJ3 Mouse Monoclonal Antibody [clone: OTI3E11]

Supplier: OriGene
ORIGTA504152EA 621 EUR
ORIGTA504152
Anti-KCNJ3 Mouse Monoclonal Antibody [clone: OTI3E11]
Antibodies
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KCNJ3 (potassium inwardly-rectifying channel, subfamily J, member 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR