Anti-UBE2J2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2535-100 ORB2535-200
BIRBORB2535-100EA 0 EUR
BIRBORB2535-100 BIRBORB2535-200
Anti-UBE2J2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: UBE2J2 (ubiquitin-conjugating enzyme E2, J2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol UBE2J2
Environmentally Preferable
Antigen name Ubiquitin-conjugating Enzyme E2, J2
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide
Form liquid
Purification purified by Protein A chromatography
Concentration 0,5 mg/ml
Storage buffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR