Anti-ADAR Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB5984-100 ORB5984-200
BIRBORB5984-100EA 0 EUR
BIRBORB5984-100 BIRBORB5984-200
Anti-ADAR Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ADAR (adenosine deaminase, RNA-specific)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human