Anti-GPC3 Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB10735-100 ORB10735-200
BIRBORB10735-100EA 0 EUR
BIRBORB10735-100 BIRBORB10735-200
Anti-GPC3 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: GPC3 (glypican 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat