Anti-HLA-DR Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB12150-100 ORB12150-500
BIRBORB12150-100EA 0 EUR
BIRBORB12150-100 BIRBORB12150-500
Anti-HLA-DR Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR