Anti-VCAN Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB13754-100 ORB13754-200
BIRBORB13754-100EA 0 EUR
BIRBORB13754-100 BIRBORB13754-200
Anti-VCAN Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: VCAN (versican)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now