Anti-PLP Mouse Monoclonal Antibody
Supplier: Biorbyt
ORB22901-1 ORB22901-500
BIRBORB22901-1EA 0 EUR
BIRBORB22901-1 BIRBORB22901-500
Anti-PLP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PLP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human