Anti-EPCAM Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB49075-100 ORB49075-200
BIRBORB49075-100EA 0 EUR
BIRBORB49075-100 BIRBORB49075-200
Anti-EPCAM Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EPCAM (epithelial cell adhesion molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR