Anti-NCF2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB49076-100 ORB49076-200
BIRBORB49076-100EA 0 EUR
BIRBORB49076-100 BIRBORB49076-200
Anti-NCF2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NCF2 (neutrophil cytosolic factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR