Anti-NLE1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126040-100 ORB126040-200
BIRBORB126040-100EA 0 EUR
BIRBORB126040-100 BIRBORB126040-200
Anti-NLE1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NLE1 (notchless homolog 1 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR