Anti-RHOD Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126308-100 ORB126308-200
BIRBORB126308-100EA 0 EUR
BIRBORB126308-100 BIRBORB126308-200
Anti-RHOD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RHOD (ras homolog family member D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR