Anti-GCFC2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126420-100 ORB126420-200
BIRBORB126420-100EA 0 EUR
BIRBORB126420-100 BIRBORB126420-200
Anti-GCFC2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GCFC2 (GC-rich sequence DNA-binding factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR