Anti-TEK Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126679-100 ORB126679-200
BIRBORB126679-100EA 0 EUR
BIRBORB126679-100 BIRBORB126679-200
Anti-TEK Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR