Anti-MRPL24 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128393-100 ORB128393-200
BIRBORB128393-100EA 0 EUR
BIRBORB128393-100 BIRBORB128393-200
Anti-MRPL24 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MRPL24 (mitochondrial ribosomal protein L24)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR