Anti-PPP2R5D Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128567-100 ORB128567-200
BIRBORB128567-100EA 0 EUR
BIRBORB128567-100 BIRBORB128567-200
Anti-PPP2R5D Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP2R5D (protein phosphatase 2, regulatory subunit B', delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR