Anti-CPT2 Rabbit Polyclonal Antibody
Supplier: Biorbyt
ORB135308-100 ORB135308-200 ORB135308-50
BIRBORB135308-100EA 0 EUR
BIRBORB135308-100 BIRBORB135308-200 BIRBORB135308-50
Anti-CPT2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CPT2 (carnitine palmitoyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat