Anti-KRT5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB135811-100 ORB135811-200 ORB135811-50
BIRBORB135811-100EA 0 EUR
BIRBORB135811-100 BIRBORB135811-200 BIRBORB135811-50
Anti-KRT5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR