Anti-TLR4 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: HTA125]
Supplier: Abcam
ABCAAB8378-100EA 719 EUR
ABCAAB8378-100
Anti-TLR4 Mouse Monoclonal Antibody FITC (Fluorescein) [clone: HTA125]
Antibodies
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: Monoclonal
Clone: HTA125
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now