Anti-SSEA1 Mouse Monoclonal Antibody [clone: MC480]

Supplier: Abcam
ABCAAB16285-200EA 860 EUR
ABCAAB16285-200
Anti-SSEA1 Mouse Monoclonal Antibody [clone: MC480]
Type: Primary
Antigen: SSEA1
Clonality: monoclonal
Clone: MC480
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR