Anti-CCND1 Rabbit Monoclonal Antibody [clone: SP4]

Supplier: Abcam
ABCAAB16663-500EA 983 EUR
ABCAAB16663-500
Anti-CCND1 Rabbit Monoclonal Antibody [clone: SP4]
Antibodies
Type: Primary
Antigen: CCND1 (cyclin D1)
Clonality: Monoclonal
Clone: SP4
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR