Anti-TNNT1 Mouse Polyclonal Antibody

Supplier: Abcam
ABCAAB89641-50EA 776 EUR
ABCAAB89641-50
Anti-TNNT1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNNT1 (troponin T type 1 (skeletal, slow))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR