Anti-MHC Class II IA + IE Rat Monoclonal Antibody FITC (Fluorescein) [clone: M5/114.15.2]

Supplier: Abcam
ABCAAB93561-100EA 744 EUR
ABCAAB93561-100
Anti-MHC Class II IA + IE Rat Monoclonal Antibody FITC (Fluorescein) [clone: M5/114.15.2]
Antibodies
Type: Primary
Antigen: MHC Class II IA + IE
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR