Anti-Bcl2 alpha Rabbit monoclonal antibody [clone: SP66]

Supplier: Abcam
ABCAAB93884-100EA 836 EUR
ABCAAB93884-100
Anti-Bcl2 alpha Rabbit monoclonal antibody [clone: SP66]
Antibodies
Type: Primary
Antigen: Bcl2 alpha
Clonality: monoclonal
Clone: SP66
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR