Anti-KCNT1 Mouse Monoclonal Antibody [clone: S3-26]

Supplier: Abcam
ABCAAB94578-100EA 821 EUR
ABCAAB94578-100
Anti-KCNT1 Mouse Monoclonal Antibody [clone: S3-26]
Antibodies
Type: Primary
Antigen: KCNT1 (potassium channel, subfamily T, member 1)
Clonality: Monoclonal
Clone: S3-26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR