Anti-MAP1A Rabbit Polyclonal Antibody

Supplier: Abcam
ABCAAB101224-100EA 719 EUR
ABCAAB101224-100
Anti-MAP1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP1A (microtubule-associated protein 1A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR