Anti-ADA Rabbit Monoclonal Antibody [clone: EPR4438]

Supplier: Abcam
AB108352-100 AB108352-1ML
ABCAAB108352-100EA 700 EUR
ABCAAB108352-100 ABCAAB108352-1ML
Anti-ADA Rabbit Monoclonal Antibody [clone: EPR4438]
Antibodies
Upozornení:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: ADA (adenosine deaminase)
Clonality: monoclonal
Clone: EPR4438
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR